Skip to Content
Merck
All Photos(1)

Key Documents

R0509

Worthington Accessories

cast aluminum roller base with 5 wheels for 35 L Dewar

Synonym(s):

dewar base

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
R$2,303.00

R$2,303.00


Estimated to ship onJune 02, 2025



Select a Size

Change View
1 EA
R$2,303.00

About This Item

UNSPSC Code:
41103000
NACRES:
NB.22

R$2,303.00


Estimated to ship onJune 02, 2025


packaging

pack of 1 ea

manufacturer/tradename

Worthington 366783

technique(s)

cryopreservation: suitable

Looking for similar products? Visit Product Comparison Guide

Caution

Weight of filled 35L Dewar can exceed 100lbs (45kg). Use of roller base is strongly recommended for safe transport.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service