Skip to Content
Merck
All Photos(1)

Key Documents

R0384

Worthington Accessories

5 shelf rack for LS750 Dewar

Synonym(s):

dewar rack

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
R$1,055.00

R$1,055.00


Available to ship onApril 07, 2025Details



Select a Size

Change View
1 EA
R$1,055.00

About This Item

UNSPSC Code:
41122806
NACRES:
NB.22

R$1,055.00


Available to ship onApril 07, 2025Details


feature

Holds 5 x boxes

packaging

pack of 1 ea

manufacturer/tradename

Worthington R036-9C34

technique(s)

cryopreservation: suitable

Looking for similar products? Visit Product Comparison Guide

General description

Each shelf holds one 3 × 3 × 2 in. (7.6 × 7.6 × 5.1cm) box with a capacity of 25 x 2mL vials. A spring-action retaining clip holds each box in place. Boxes not included.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service