Skip to Content
Merck
All Photos(1)

Key Documents

PZ0310

Sigma-Aldrich

PF-04995274

≥98% (HPLC)

Synonym(s):

4-{4-[4-Tetrahydrofuran-3-yloxy)-benzo[d]isoxazol-3-yloxymethyl]-piperidin-1-ylmethyl}-tetrahydropyran-4-ol, PF-4995274, PF04995274, TBPT, Tetrahydro-4-[[4-[[[4-[[(3R)-tetrahydro-3-furanyl]oxy]-1,2-benzisoxazol-3-yl]oxy]methyl]-1-piperidinyl]methyl]-2H-pyran-4-ol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$716.00
25 MG
R$2,881.00

R$716.00


Estimated to ship onApril 13, 2025



Select a Size

Change View
5 MG
R$716.00
25 MG
R$2,881.00

About This Item

Empirical Formula (Hill Notation):
C23H32N2O6
CAS Number:
Molecular Weight:
432.51
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

R$716.00


Estimated to ship onApril 13, 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

OC1(CCOCC1)CN(CC2)CCC2COC3=NOC4=C3C(O[C@@H]5CCOC5)=CC=C4

InChI

1S/C23H32N2O6/c26-23(7-12-27-13-8-23)16-25-9-4-17(5-10-25)14-29-22-21-19(30-18-6-11-28-15-18)2-1-3-20(21)31-24-22/h1-3,17-18,26H,4-16H2/t18-/m1/s1

InChI key

WLLOFQROROXOMO-GOSISDBHSA-N

Biochem/physiol Actions

PF-04995274 favors the production of cyclic adenosine monophosphate (cAMP).[1]
PF-04995274 is a potent partial agonist of serotonin 5HT4 receptor with Ki = 0.15 - 0.46 nM for 5-HT4 isoforms a, b, d and e. 5-HT4 agonists modulate cholinergic function are being studied as possible treatment for cognitive disorders associated with Alzheimer′s Disease. PF-04995274 is brain penetrant and was shown to increase brain acetylcholine levels and reversed scopolamine-induced learning deficits in the rat Morris water maze test.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael A Brodney et al.
Journal of medicinal chemistry, 55(21), 9240-9254 (2012-09-15)
The cognitive impairments observed in Alzheimer's disease (AD) are in part a consequence of reduced acetylcholine (ACh) levels resulting from a loss of cholinergic neurons. Preclinically, serotonin 4 receptor (5-HT(4)) agonists are reported to modulate cholinergic function and therefore may

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service