Skip to Content
Merck
All Photos(3)

Key Documents

N4753

Sigma-Aldrich

N-(2-Nitrophenylsulfenyl)-γ-aminobutyric acid (dicyclohexylammonium) salt

powder

Synonym(s):

NPS-GABA

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
R$2,388.00

R$2,388.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Select a Size

Change View
5 G
R$2,388.00

About This Item

Empirical Formula (Hill Notation):
C10H12N2O4S · C12H23N
CAS Number:
Molecular Weight:
437.60
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26

R$2,388.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

form

powder

color

yellow

storage temp.

−20°C

SMILES string

C1CCC(CC1)NC2CCCCC2.OC(=O)CCCNSc3ccccc3[N+]([O-])=O

InChI

1S/C12H23N.C10H12N2O4S/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;13-10(14)6-3-7-11-17-9-5-2-1-4-8(9)12(15)16/h11-13H,1-10H2;1-2,4-5,11H,3,6-7H2,(H,13,14)

InChI key

DWPCVLOTBKFZQB-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service