Skip to Content
Merck
All Photos(1)

Key Documents

93662

Sigma-Aldrich

Fluorescent red NIR 700

suitable for fluorescence

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
R$3,103.00

R$3,103.00


Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
R$3,103.00

About This Item

Empirical Formula (Hill Notation):
C39H44N2O6S
Molecular Weight:
668.84
MDL number:
UNSPSC Code:
12352108
NACRES:
NA.32

R$3,103.00


Estimated to ship onJune 02, 2025


Request a Bulk Order

form

powder

solubility

DMSO: soluble

fluorescence

λex 672 nm; λem ~735 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

2-8°C

General description

Fluorescent red 700 is a fluorescent dye for the near infrared area. Extremely long wavelength fluorescence is especially well suited for applications where sample autofluorescence might be critical or penetration of tissue or other matrix is required.

Analysis Note

further Exc.: 700 nm (results in a lower emission)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service