Skip to Content
Merck
All Photos(1)

Key Documents

LVCAS9NEO

Sigma-Aldrich

Cas9 Neomycin Lenti Particles

Synonym(s):

Lenti Particles

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
R$3,098.00

R$3,098.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
R$3,098.00

About This Item

UNSPSC Code:
41106609
NACRES:
NA.51

R$3,098.00


Check Cart for Availability

Request a Bulk Order

packaging

vial of 200 μL

Quality Level

concentration

≥1 x 10 6 VP/ml virus (via p24 assay)

application(s)

CRISPR

selection

neomycin / G418

shipped in

dry ice

storage temp.

−70°C

General description

Ready-to-use Cas9 lentiviral particles are ideal for transducing a wide range of cell lines. The Cas9 Neomycin Lentiviral particles efficiently integrate Cas9 into the genomes of both dividing and non-dividing cell lines for use with CRISPR genome editing. Stably integrate Cas9 and Neo using the convenient, ready-to-use lentiviral particles while avoiding the tedious lentivirus production process.

Application

Functional Genomics/Screening /Target Validation/Genome Editing

Components

200 ul of purified lentiviral particles (minimum of 1 X 106 TU/ml via a p24 assay) expressing Cas9 only with blasticidin resistance.

Linkage

Click here to order gRNA plasmid and virus.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Genome-wide screening with optimized gRNAs per gene ensures specific and efficient knockout, controlling time and cost.

Questions

  1. Is there a protocol associated with the Cas9 Neomycin lentiparticles? I will be transfecting some glioblastoma cells with the virus and am looking for a starting point for protocol design.

    1 answer
    1. Titer values are lot specific and available in the Certificate of Analysis. Please navigate to the documentation section to view the sample Certificate:
      https://www.sigmaaldrich.com/product/sigma/lvcas9neo#product-documentation

      A specific protocol for this item is not available, however this item is required for use in several Lentiviral CRISPR Pool products. Reviewing the Technical Bulletin for these products may be helpful. Please see the link below:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/294/472/hwgcrisprbul.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service