Key Documents
HS120649
Reagent Reservoir
capacity 100 mL, polystyrene, non-sterile, pack of 100 (bulk packed)
Select a Size
100 EACH
R$629.00
R$629.00
Available to ship onApril 11, 2025Details
Select a Size
Change View
100 EACH
R$629.00
About This Item
UNSPSC Code:
41121522
NACRES:
NB.01
R$629.00
Available to ship onApril 11, 2025Details
material
V-bottom bottom
polystyrene
sterility
non-sterile
packaging
pack of 100 (bulk packed)
manufacturer/tradename
Heathrow Scientific 120649
capacity
100 mL
General description
Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures. Graduation markingson the top edge and inner walls allows measurements of liquid to be easily identified and reduces waste of expensive reagents. Pour-off spouts design on all four corners enables easy spill-free pouring and reduces chance of dripping.
Features and Benefits
  • Trough design with a “v” shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
  • Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
  • Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
  • Designed to stay flat on the benchtop and not warp
  • Sealed in easy-to-open bags. No scissors needed to open
  • Disposable
  • Design Patent Pending
Choose from one of the most recent versions:
Certificates of Analysis (COA)