This product is manufactured by Heathrow Scientific, part number 120648 (found in the "Properties" section of the product page). According to the manufacture, this product is designed for multi-channel pipettes. The interior dimensions of this product are as follows: 4.3 x 1.9 x 1.3 in (10.9 x 4.8 x 3.2 cm). More information can be found at this link:
https://www.heathrowscientific.com/reagent-reservoirs
HS120648
Reagent Reservoir
capacity 100 mL, polystyrene, sterile; electron beam irradiated, pack of 100 (individually wrapped)
Synonym(s):
Reagent Container, Reagent Storage
Select a Size
R$1,274.00
Select a Size
About This Item
R$1,274.00
Recommended Products
material
V-bottom bottom
polystyrene
sterility
sterile; electron beam irradiated
packaging
pack of 100 (individually wrapped)
manufacturer/tradename
Heathrow Scientific 120648
capacity
100 mL
General description
Features and Benefits
- Trough design with a “v” shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
- Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
- Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
- Designed to stay flat on the benchtop and not warp
- Sealed in easy-to-open bags. No scissors needed to open
- Disposable
- Design Patent Pending
Choose from one of the most recent versions:
Certificates of Analysis (COA)
-
Is this product working with multi-channel pipett? I cant find the dimentions. BR/Josefin
1 answer-
Helpful?
-