Skip to Content
Merck
All Photos(1)

Key Documents

HLP501

Sigma-Aldrich

Normal Human Intrahepatic Biliary Epithelial Cells (Cholangioctyes); passage 1

Single-Donor Cryopreserved Cholangioctyes cells isolated from human livers, high viability, and characterized by cell specific markers

Synonym(s):

Homogenous population isolated from whole human livers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106509
NACRES:
NA.81

biological source

human liver

form

frozen liquid

packaging

pkg of 1 vial (contains ≥0.25 million cells)
pkg of 1 vial (contains ≥0.5 million cells)
pkg of 1 vial (contains ≥1 million cells)

technique(s)

cell culture | mammalian: suitable

color

cloudy

storage temp.

-140 to -196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details. Primary human Cholangiocytes, also known as human intrahepatic biliary epithelial cells, are epithelial cells that line the intrahepatic bile ducts. Chonlangiocytes are important for ductal bile modification and are targeted in multiple liver diseases, such as primary biliary cirrhosis, cholangiocarcinoma and sclerosing cholangitis.

Application

Human cholangiocytes are ideal in studies of: toxicity, bile production and absorption, bile duct fibrosis, hormone signaling, cholangitis, cholangiocarcinoma and more. We offer matched sets of cholangiocytes cells from same donor as Hepatocytes, which can be used for co-culture systems to better simulate the complex microenvironment of the liver and gain insights into various liver-related diseases and conditions.

Features and Benefits

Our cryopreserved human Cholangiocytes cells are derived from the human liver, with each donor providing documented consent for research use of non-transplantable organs or tissues. Following the primary culture, these cells are cryopreserved. Each lot undergoes testing for specific cell markers and comes with a guarantee of ≥ 70% post-thaw viability. In addition, all cells have been tested for the absence of HIV, HBV, HCV, Syphillis and microbial contaminants (fungi, bacteria, and mycoplasma).

Warning

Each donor is tested and found non-reactive to viral DNA from HIV-1, HIV-2, HTLV I, HTLV II, hepatitis B and hepatitis C. However, no known test can offer complete assurance that the viruses that cause HIV-1, HIV-2, HTLV I, HTLV II, hepatitis B and hepatitis C are not present. Proper precautions and biological containment should be taken when handling cells of human origin, due to their potential biohazardous nature. All human based products should be handled at a BSL-2 (Biosafety Level 2) or higher.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Explore answers to frequently asked questions (FAQs) about culturing primary human hepatocytes and other liver cells including Stellate cells, Kupffer cells, Sinusoidal Endothelial cells, and more.

Protocols

Discover our collection of primary human hepatic Kupffer cells and protocol for thawing, plating, and growing Kupffer cells. Find the formulation for Kupffer culture maintenance media.

Discover our collection of primary human hepatic sinusoidal endothelial cells, a protocol for thawing, plating, and growing HHSECs, and a recipe for our HHSEC maintenance media formulation.

Related Content

Explore our primary liver cells collection, including primary human Hepatocytes (PHH), spheroid-qualified Hepatocytes, Kupffer cells, Stellate cells, Cholangiocytes, and Sinusoidal Endothelial cells.

Discover our collection of primary human hepatic stellate cells and protocol for thawing, plating, and growing stellate cells. Find our stellate culture maintenance media formulation.

Discover our primary human intrahepatic biliary epithelial cells, also known as cholangiocytes. Explore our plating and maintenance media formulations and protocol for culture.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service