MilliporeSigma
Your New Hepatocyte Partner
Ready-to-screen and validated Human primary Hepatocytes, Stellate cells, Kupffer cells, and more.
Pre-tested, Cryopreserved, and Ready-to-Screen Human Primary Liver Cells
Male scientist sitting at the tissue culture hood holding a handheld pipette gun pipetting red media into a six well cell culture plate. Three media bottles are in the background and a T flask with media is in the foreground on a Millicell® DCI.
We now offer a wide range of cryopreserved, ready-to-screen normal human primary liver cells. Our extensive range of primary human hepatocytes includes suspension hepatocytes from single and pooled donors, as well as plateable hepatocytes and spheroid-qualified hepatocytes for 3D cell culture applications. Our pooled suspension hepatocytes are normalized for cell number to ensure that each donor contributes an equal contribution to the pool.
Additionally, we offer various other liver cells including Stellate cells, Kupffer cells, Cholangiocytes, and Sinusoidal Endothelial cells. Our primary liver cells are assessed for yield, viability, and morphology and are characterized by attachment, surface/internal marker expression, and other cell-specific methods.
Phase contrast microscopy image of stellate cells. Light grey square with dark grey dots and elongated dark grey rectangles representing the cells.
Figure 1.Stellate cells after 4 days, imaged with the Millicell® DCI Digital Cell Imager at 10x magnification setting. The cells display an elongated shape characteristic to Stellate cells.
Our collection of liver primary cells is directly harvested from whole human livers and can be used to model the physiology and function of livers in vitro. The portfolio includes primary human Hepatocytes, Stellate cells, Kupffer cells, Cholangiocytes, and Sinusoidal endothelial cells. Our liver primary cell collection has high post‐thaw viability and yield, as well as the following key advantages:
  • Increased reproducibility – we provide large lot sizes of the same donor so they can be used for long-term studies.
  • Cells are cryopreserved – increasing the ease of use.
  • Extensive characterization – marker expression, induction and metabolic assays, morphology and attachment.
  • Multi-parameter donor demographics and lot specific orders.
  • Full documentation - cells are ethically sourced to ensure full regulatory compliance.
What human liver cells are offered? Are they ethically collected? Are they screened? Get answers to these questions and more with our Hepatocytes frequently asked questions technical article.
Related Products
Sorry, an unexpected error has occurred
Network error: Failed to fetch
Related Resources
Page 1 of 2