Skip to Content
Merck
All Photos(1)

Key Documents

EP5702734004

Eppendorf® Adapter for 85/100 mL Round Bucket

holds Vacutainer and Falcon tubes 50 ml, pack of 2 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103911
NACRES:
NB.82
Pricing and availability is not currently available.

feature

holds Vacutainer and Falcon tubes 50 ml

packaging

pack of 2 ea

manufacturer/tradename

Eppendorf® 5702734004

technique(s)

centrifugation: suitable

compatibility

for use with 100 mL round buckets

Looking for similar products? Visit Product Comparison Guide

General description

adapter for use with Eppendorf 85/100 mL round bucket

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service