Skip to Content
Merck
All Photos(1)

Key Documents

CLS430236

Corning® large volume centrifuge tube support cushions

Synonym(s):

centrifuge tube holders, centrifuge tube support cushions, centrifuge tube supports

Sign Into View Organizational & Contract Pricing

Select a Size

6 EA
R$460.00

R$460.00


Check Cart for Availability


Select a Size

Change View
6 EA
R$460.00

About This Item

UNSPSC Code:
41103911
NACRES:
NB.14

R$460.00


Check Cart for Availability

material

polyetherimide

sterility

non-sterile

feature

reusable
Holds 250 mL tubes

packaging

pack of 1
case of 6

manufacturer/tradename

Corning 430236

technique(s)

centrifugation: suitable

color

gray

Looking for similar products? Visit Product Comparison Guide

General description

Corning® centrifuge tube cushions are manufactured from polyetherimide. They provide the required support and prevent large-capacity centrifuge tubes from collapsing during centrifugation.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service