Skip to Content
Merck
All Photos(1)

Documents

EP022639404

Rotor for Eppendorf® 5702/5702R/5702RH Centrifuges

to hold, 30 x 15 mL vessels

Synonym(s):

Rotor for Eppendorf® 5702/5702R/5702RH Centrifuges

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103909

feature

to hold 30 x 15 mL vessels

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 22639404

parameter

4,400 max. RCF (2,750 xg)

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models 5702/5702R/5702RH

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Eppendorf fixed angle rotor acccommodates up to thirty 15mL Falcon centrifuges tubes. Includes 30 adapters (F-35-30-17) and 30 rubber mats.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service