Skip to Content
Merck
All Photos(1)

Key Documents

D7946

Sigma-Aldrich

DIM-C-pPhOCH3

≥98% (HPLC)

Synonym(s):

1,1-bis(3′-Indolyl)-1-(p-methoxyphenyl)methane; 3,3′-[(4-Methoxyphenyl)methylene]bis-1H-indole

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$1,043.00
25 MG
R$4,243.00

R$1,043.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
R$1,043.00
25 MG
R$4,243.00

About This Item

Empirical Formula (Hill Notation):
C24H20N2O
CAS Number:
Molecular Weight:
352.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

R$1,043.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

off-white to light yellow

solubility

DMSO: ≥30 mg/mL

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C(c2c[nH]c3ccccc23)c4c[nH]c5ccccc45

InChI

1S/C24H20N2O/c1-27-17-12-10-16(11-13-17)24(20-14-25-22-8-4-2-6-18(20)22)21-15-26-23-9-5-3-7-19(21)23/h2-15,24-26H,1H3

InChI key

ZCCMKJAXOIFTHH-UHFFFAOYSA-N

Biochem/physiol Actions

DIM-C-pPhOCH3 is a Nerve Growth Factor-Induced Bα (NGFI-Bα, Nur77) agonist

Features and Benefits

This compound is featured on the Nuclear Receptors (Non-Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katriana A Popichak et al.
Molecular pharmacology, 94(4), 1174-1186 (2018-08-17)
Inflammatory activation of glial cells promotes loss of dopaminergic neurons in Parkinson disease. The transcription factor nuclear factor κB (NF-κB) regulates the expression of multiple neuroinflammatory cytokines and chemokines in activated glial cells that are damaging to neurons. Thus, inhibition

Articles

We offer many products related to non-steroid nuclear receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service