CLS431840
Corning® centrifuge bottle
with silicon O-ring screw cap, capacity 250 mL, polypropylene
Synonym(s):
centrifugation bottles, digestion bottles, shaking bottles
Select a Size
R$2,954.00
Select a Size
About This Item
R$2,954.00
Recommended Products
material
flat bottom tube
polypropylene
round bottle
translucent bottle
wide-mouth bottle
sterility
non-sterile
feature
autoclavable
cap
packaging
pkg of 36 ea
manufacturer/tradename
Corning 431840
parameter
27500 × g max. RCF
capacity
250 mL
Related Categories
Features and Benefits
- Corning® centrifuge bottles have a specially designed cap for improved grip
- Includes clear graduations and textured marking spot.
- Bottles are designed with extra-thick walls for high durability
- Wide mouth for easier liquid filling
- Compatible with most centrifuges and do not require special buckets
- Unique design aid researchers to reduce the number of centrifugation cycles and enhance productivity
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)