CLS11110
Corning® CellSTACK® accessories
universal cap with vented overcap, 2-stack, tissue-culture treated, pack of 4 ea
Synonym(s):
Corning cap
About This Item
Recommended Products
packaging
pack of 4 ea
manufacturer/tradename
Corning 11110
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service