Skip to Content
Merck
All Photos(1)

Key Documents

M-MSV-BALB/3T3

90030802, mouse embryo, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

M-MSV-BALB/3T3, 90030802, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

biological source

mouse embryo

packaging

tube of 5 μg 90030802-DNA-5UG
pkg of vial of cells 90030802-1VL

growth mode

Adherent

karyotype

Modal no. 57

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse BALB/3T3 embryo, Moloney sarcoma virus transformed

Cell Line Description

Derived from BALB/3T3 clone A31 (Sigma Catalogue number. 86110401) by infection with M-MSV. A M-MSV non-producer, but addition of MuLV permits rescue of the MSV genome. Cells produce tumours in suitable hosts; lack contact inhibition in vitro cultures; morphologically identical to virus releasing lines.

Application

Transformation and virus studies: M-MSV, MuLV

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:10 i.e. seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service