Skip to Content
Merck
All Photos(1)

Key Documents

GH1

89072610, rat brain, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

GH1, 89072610

biological source

rat brain

packaging

tube of 5 μg 89072610-DNA-5UG
pkg of vial of cells 89072610-1VL

growth mode

Semi-adherent

karyotype

Hypodiploid, modal no. 40.

morphology

Epithelial

products

Rat growth hormone, lactogenic hormones.

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat Wistar-Furth pituitary tumour

Cell Line Description

Derived from pituitary gland of Wistar-Furth rats. The cells secrete growth hormone at a rate of 1μg/ml cell protein/day. Re-inoculation of cloned cells back to Wistar-Furth rats results in a marked increase in body weight and stimulation of linear growth in both intact and hypophysectomized animals independent of rat tumour size.

Application

Hormone studies

Culture Medium

Ham′s F10 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:3 i.e. seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C. These cells only semi-attach and may return to suspension during transit. If this has occurred on receipt of the cells pellet cells b

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service