Skip to Content
Merck
All Photos(1)

Documents

CL 21

86080602, Human x Mouse Hybridoma unknown/unspecified, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

CL 21, 86080602

biological source

Human x Mouse Hybridoma unknown/unspecified

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse / Human hybrid, SV-40 transformed

Cell Line Description

The cell line CL21 has been established by hybridisation of C57 BL mouse peritoneal macrophages with SV40 transformed human skin fibroblasts. The human cells were derived from a patient with Lesch-Nyhan syndrome and are deficient in hypoxanthine guanine phosphoribosyltransferase (HGPRT). They have been reported to contain the human chromosome 7 and could also contain subchromosomal fragments of chromosome 17.

Application

Somatic cell hybrid mapping

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service