Recommended Products
product name
C127I, 90060504
biological source
mouse breast
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Related Categories
Cell Line Origin
Mouse RIII mammary tumour
Cell Line Description
Non-transformed clone from a RIII mouse. Supernatant fluid is negative for RT activity. Useful for visualisation of sarcoma virus-induced foci, in vitro quantitative assay for bovine papilloma virus. Suitable host for transformation with bovine papilloma virus DNA plasmids.
Application
Virus and transformation studies: Sarcoma virus, Bovine papilloma virus
Culture Medium
DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:5 to 1:15 i.e. seeding at 1-2x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service