C4743
Monoclonal Anti-Cytochrome P450 3A5 antibody produced in mouse
~2 mg/mL, clone F18 P3 B6, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Anti-CP35, Anti-CYP3A5, Anti-P450PCN3, Anti-PCN3
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
F18 P3 B6, monoclonal
form
buffered aqueous solution
mol wt
antigen ~55 kDa
species reactivity
human
concentration
~2 mg/mL
technique(s)
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: 0.5-1.0 μg/mL using human recombinant cytochrome P450 3A5.
isotype
IgG1
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... CYP3A5(1577)
Specificity
Immunogen
Application
- immunohistochemistry
- indirect ELISA
- western blotting at a concentration of 0.5-1.0 μg/mL using human recombinant cytochrome P450 3A5
Biochem/physiol Actions
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service