Skip to Content
Merck
All Photos(1)

Key Documents

C3305

Sigma-Aldrich

Carbacyclin

≥90% (HPLC)

Synonym(s):

6a-Carba-prostaglandin I2, Carbocyclic PGI2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H34O4
CAS Number:
Molecular Weight:
350.49
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

≥90% (HPLC)

storage temp.

−20°C

SMILES string

[H][C@]12C[C@@H](O)[C@H](\C=C\[C@@H](O)CCCCC)[C@@]1([H])CC(\C2)=C\CCCC(O)=O

InChI

1S/C21H34O4/c1-2-3-4-8-17(22)10-11-18-19-13-15(7-5-6-9-21(24)25)12-16(19)14-20(18)23/h7,10-11,16-20,22-23H,2-6,8-9,12-14H2,1H3,(H,24,25)/b11-10+,15-7+/t16-,17-,18+,19-,20+/m0/s1

InChI key

XZFRIPGNUQRGPI-WBQKLGIQSA-N

Gene Information

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chuan-Ming Hao et al.
The Journal of biological chemistry, 277(24), 21341-21345 (2002-04-03)
COX2-selective non-steroidal anti-inflammatory drugs (NSAIDs) cause selective apoptosis of renal medullary interstitial cells (RMIC) in vivo and reduce their ability to tolerate hypertonic stress in vitro. To determine the mechanism by which COX2 activity promotes RMIC viability, we examined the
R A Gupta et al.
Proceedings of the National Academy of Sciences of the United States of America, 97(24), 13275-13280 (2000-11-23)
There is evidence from both genetic and pharmacologic studies to suggest that the cyclooxygenase-2 (COX-2) enzyme plays a causal role in the development of colorectal cancer. However, little is known about the identity or role of the eicosanoid receptor pathways
Charlotte C Jouvene et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(12), 13794-13807 (2019-10-08)
Resolution of inflammation is an active process regulated by specialized proresolving mediators where we identified 3 new pathways producing allylic epoxide-derived mediators that stimulate regeneration [i.e., peptido-conjugates in tissue regeneration (CTRs)]. Here, using self-limited Escherichia coli peritonitis in mice, we
M J Kim et al.
Molecular genetics and metabolism, 74(3), 362-369 (2001-11-16)
We have found that sebaceous epithelial cell (sebocyte) differentiation is induced by cognate ligand-agonists of either peroxisome proliferator-activated receptors (PPARs) or retinoid X receptors (RXRs). In this study, we tested the hypothesis that PPAR-RXR cooperation is used in sebocytes as
A C Maurin et al.
Calcified tissue international, 76(5), 385-392 (2005-05-04)
As previously reported, the age-related association between bone loss and increased marrow adipose volume may involve inhibitory effects of polyunsaturated fatty acids (PUFAs) potentially released by medullary adipocytes on osteoblastic proliferation and cell cycle progression. Because PUFAs have been reported

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service