Skip to Content
Merck
All Photos(1)

Key Documents

A7604

Sigma-Aldrich

Atracurium besylate

≥98% (HPLC), powder

Synonym(s):

2,2′-[1,5-Pentanediylbis[oxy(3-oxo-3,1-propanediyl)]]bis[1-[3,4-dimethoxyphenyl)methyl]-1,2,3,4-tetrahydro-6,7-dimethoxy-2-methylisoquinolinium]dibenzenesulfonate, BW-33A, Tracrium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C65H82N2O18S2
CAS Number:
Molecular Weight:
1243.48
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

color

white

solubility

H2O: ~27 mg/mL

storage temp.

2-8°C

SMILES string

[O-]S(=O)(=O)c1ccccc1.[O-]S(=O)(=O)c2ccccc2.COc3ccc(CC4c5cc(OC)c(OC)cc5CC[N+]4(C)CCC(=O)OCCCCCOC(=O)CC[N+]6(C)CCc7cc(OC)c(OC)cc7C6Cc8ccc(OC)c(OC)c8)cc3OC

InChI

1S/C53H72N2O12.2C6H6O3S/c1-54(22-18-38-32-48(62-7)50(64-9)34-40(38)42(54)28-36-14-16-44(58-3)46(30-36)60-5)24-20-52(56)66-26-12-11-13-27-67-53(57)21-25-55(2)23-19-39-33-49(63-8)51(65-10)35-41(39)43(55)29-37-15-17-45(59-4)47(31-37)61-6;2*7-10(8,9)6-4-2-1-3-5-6/h14-17,30-35,42-43H,11-13,18-29H2,1-10H3;2*1-5H,(H,7,8,9)/q+2;;/p-2

InChI key

XXZSQOVSEBAPGS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Nicotinic acetylcholine receptor antagonist.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malin Jonsson et al.
European journal of pharmacology, 497(2), 173-180 (2004-08-13)
Neuromuscular blocking agents predominantly block muscle type nicotinic acetylcholine receptors as opposed to the neuronal type. However, there is growing evidence that neuromuscular blocking agents have affinity to some neuronal nicotinic acetylcholine receptors. The carotid body chemoreceptor as the essential
Arnaud Louvet et al.
Veterinary radiology & ultrasound : the official journal of the American College of Veterinary Radiology and the International Veterinary Radiology Association, 56(3), 286-295 (2014-12-23)
Early detection of liver metastases may improve the prognosis for successful treatment in dogs with primary tumors. Hepatobiliary-specific contrast agents have been shown to allow an increase in magnetic resonance imaging (MRI) detection of liver metastases in humans. The purpose
Gudrun Schöffmann et al.
Veterinary anaesthesia and analgesia, 39(3), 256-265 (2012-03-17)
To determine the effects of age, sevoflurane and isoflurane on atracurium-induced neuromuscular blockade in 3-16 week-old lambs. Prospective randomized experimental trial. Twenty-six Scottish blackface ewe-lambs were anaesthetized for spinal surgery when either 3-6 (mean age 4.6 weeks; n = 18)
Maxime Gauberti et al.
Cerebrovascular diseases (Basel, Switzerland), 33(4), 329-339 (2012-02-22)
The lack of a relevant stroke model in large nonhuman primates hinders the development of innovative diagnostic/therapeutic approaches concerned with this cerebrovascular disease. Our objective was to develop a novel and clinically relevant model of embolic stroke in the anesthetized
Michele Carron et al.
Anesthesiology, 117(2), 309-320 (2012-05-23)
The stress responses from tracheal intubation are potentially dangerous in patients with higher cardiovascular risk, such as obese patients. The primary outcome objective of this study was to test whether, in comparison with the endotracheal tube (ETT), the Proseal™ Laryngeal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service