A2793
Arg-Arg-Arg-Glu-Glu-Glu-Thr-Glu-Glu-Glu
lyophilized powder
Select a Size
R$1,426.00
Estimated to ship onMay 22, 2025
Select a Size
About This Item
R$1,426.00
Estimated to ship onMay 22, 2025
Recommended Products
form
lyophilized powder
storage temp.
−20°C
SMILES string
C[C@@H](O)[C@H](NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](N)CCCNC(N)=N)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCC(O)=O)C(O)=O
InChI
1S/C52H87N19O24/c1-23(72)39(48(93)69-30(11-17-36(79)80)45(90)67-29(10-16-35(77)78)46(91)70-32(49(94)95)13-19-38(83)84)71-47(92)31(12-18-37(81)82)68-44(89)28(9-15-34(75)76)66-43(88)27(8-14-33(73)74)65-42(87)26(7-4-22-62-52(58)59)64-41(86)25(6-3-21-61-51(56)57)63-40(85)24(53)5-2-20-60-50(54)55/h23-32,39,72H,2-22,53H2,1H3,(H,63,85)(H,64,86)(H,65,87)(H,66,88)(H,67,90)(H,68,89)(H,69,93)(H,70,91)(H,71,92)(H,73,74)(H,75,76)(H,77,78)(H,79,80)(H,81,82)(H,83,84)(H,94,95)(H4,54,55,60)(H4,56,57,61)(H4,58,59,62)/t23-,24+,25+,26+,27+,28+,29+,30+,31+,32+,39+/m1/s1
InChI key
FYEPIIASCNUBEH-MJBCRNAASA-N
Gene Information
human ... CSNK2A1(1457) , CSNK2A2(1459) , CSNK2B(1460)
mouse ... CSNK2A1(12995) , CSNK2A2(13000) , CSNK2B(13001)
rat ... CSNK2A1(116549) , CSNK2B(81650)
Amino Acid Sequence
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service