Skip to Content
Merck
All Photos(1)

Key Documents

94121

Sigma-Aldrich

Baicalin

≥99.0% (HPLC)

Synonym(s):

5,6-Dihydroxy-4-oxo-2-phenyl-4H-1-benzopyran-7-yl-β-D-glucopyranosiduronic acid, Baicalein 7-O-β-D-glucuronide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18O11
CAS Number:
Molecular Weight:
446.36
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

biological source

synthetic

Assay

≥99.0% (HPLC)

form

solid

optical activity

[α]/D -83±3°, c = 1 in DMSO

technique(s)

HPLC: suitable

mp

202-205 °C (dec.) (lit.)

SMILES string

O[C@@H]1[C@@H](O)[C@@H](O[C@@H]([C@H]1O)C(O)=O)Oc2cc3OC(=CC(=O)c3c(O)c2O)c4ccccc4

InChI

1S/C21H18O11/c22-9-6-10(8-4-2-1-3-5-8)30-11-7-12(14(23)15(24)13(9)11)31-21-18(27)16(25)17(26)19(32-21)20(28)29/h1-7,16-19,21,23-27H,(H,28,29)/t16-,17-,18+,19-,21+/m0/s1

InChI key

IKIIZLYTISPENI-ZFORQUDYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Baicalin, a flavonoid compound with potential therapeutic value, is used to study its anti-inflammatory and anti-oxidant properties. Baicalin is also used to study its potential as an anti-sepsis agent.

Biochem/physiol Actions

Baicalin, a component of Traditional Chinese Medicine (TCM), is a flavonoid component found in several species in the genus Scutellaria. Baicalin is an inhibitor of prolyl endopeptidases.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T B Ng et al.
Life sciences, 66(8), 709-723 (2000-02-19)
A variety of flavonoids, lignans, an alkaloid, a bisbenzyl, coumarins and terpenes isolated from Chinese herbs was tested for antioxidant activity as reflected in the ability to inhibit lipid peroxidation in rat brain and kidney homogenates and rat erythrocyte hemolysis.
Zhenhua Pei et al.
Journal of periodontology, 85(9), 1283-1290 (2014-01-31)
Periodontitis is the most common cause of tooth loss in adults. Periodontal ligament cell (PLC)-based therapy is considered one of the most promising methods in periodontal tissue regeneration. The traditional Chinese medicine baicalin has been shown to possess antimicrobial and
Z Gao et al.
Biochimica et biophysica acta, 1472(3), 643-650 (1999-11-24)
Free radical scavenging and antioxidant activities of baicalein, baicalin, wogonin and wogonoside, the four major flavonoids in the radix of Scutellaria baicalensis Georgi, were examined in different systems. ESR results showed that baicalein and baicalin scavenged hydroxyl radical, DPPH radical
Xi-Ping Zhang et al.
World journal of gastroenterology, 13(38), 5079-5089 (2007-09-19)
To investigate the protective effects and mechanisms of Baicalin and octreotide on renal injury of rats with severe acute pancreatitis (SAP). One hundred and eighty SD rats were randomly assigned to the model group, Baicalin-treated group, octreotide-treated group and sham
Miao Lin et al.
BMC complementary and alternative medicine, 14, 19-19 (2014-01-15)
Renal ischemia-reperfusion injury (IRI) increases the rates of acute kidney failure, delayed graft function, and early mortality after kidney transplantation. The pathophysiology involved includes oxidative stress, mitochondrial dysfunction, and immune-mediated injury. The anti-oxidation, anti-apoptosis, and anti-inflammation properties of baicalin, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service