Skip to Content
Merck
All Photos(1)

Key Documents

D1700000

Diethylstilbestrol monomethyl ether

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Mestilbol, NSC 16971

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22O2
CAS Number:
Molecular Weight:
282.38
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

diethylstilbestrol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O(C)c1ccc(cc1)\C(=C(/CC)\c2ccc(cc2)O)\CC

InChI

1S/C19H22O2/c1-4-18(14-6-10-16(20)11-7-14)19(5-2)15-8-12-17(21-3)13-9-15/h6-13,20H,4-5H2,1-3H3/b19-18+

InChI key

CKMDPZMWUZDKAI-VHEBQXMUSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Diethylstilbestrol monomethyl ether EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Sakakibara et al.
Mutation research, 263(4), 269-276 (1991-08-01)
We previously reported that diethylstibestrol (DES) and its derivatives inhibit the in vitro polymerization of microtubule proteins isolated from porcine brain (Sato et al., 1987). We found that the presence of the free hydroxy group of DES was indispensable for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service