Skip to Content
Merck
All Photos(1)

Key Documents

T9073

Supelco

1-(α,α,α-Trifluoro-m-tolyl)piperazine

analytical standard, 96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13F3N2
CAS Number:
Molecular Weight:
230.23
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Assay

96%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.521 (lit.)

density

1.226 g/mL at 25 °C (lit.)

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

FC(F)(F)c1cccc(c1)N2CCNCC2

InChI

1S/C11H13F3N2/c12-11(13,14)9-2-1-3-10(8-9)16-6-4-15-5-7-16/h1-3,8,15H,4-7H2

InChI key

KKIMDKMETPPURN-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Serotonin receptor agonist with preference for 5-HT1B serotonin receptors

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barry K Logan et al.
Journal of analytical toxicology, 34(9), 587-589 (2010-11-16)
A series of patients whose urine screened positive for 3,4-methylenedioxymethamphetamine (MDMA) using a commercial enzyme immunoassay test (Ecstasy EMIT II assay), failed to confirm by substance-specific liquid chromatography-tandem mass spectrometry tests for MDMA. Further evaluation of these urine specimens indicates
Ustana Antia et al.
The New Zealand medical journal, 122(1307), 55-68 (2010-02-12)
BZP and TFMPP are amphetamine-like recreational drugs and the major active components of 'party pills'. The pharmacodynamic effects of these neurally active drugs are thought to be dependent on their activity at DA and 5-HT receptors and several studies report
Piperazines: a new class of drug of abuse has landed in Hong Kong.
W T Poon et al.
Hong Kong medical journal = Xianggang yi xue za zhi, 16(1), 76-77 (2010-02-04)
Mitsuhiro Wada et al.
Biomedical chromatography : BMC, 26(1), 21-25 (2011-03-10)
An HPLC-fluorescence detection method for simultaneous determination of N-benzylpiperazine (BZP) and 1-(3-trifluoromethylphenyl)piperazine (TFMPP) labeled with 4-(4,5-diphenyl-1 H-imidazol-2-yl)benzoyl chloride (DIB-Cl) was described. DIB-BZP and -TFMPP were well separated within 13 min without interference of peaks from plasma components. The lower detection
Ushtana Antia et al.
The New Zealand medical journal, 122(1303), 29-38 (2009-10-24)
This study explores potential drug-drug interactions between BZP and TFMPP. This was achieved by comparing the metabolism and pharmacokinetics of BZP and TFMPP when taken together with previously published data on their individual metabolism and pharmacokinetics. Blood and urine samples

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service