SML2058
5,7-Dihydroxytryptamine hydrobromide
≥94% (HPLC)
Synonym(s):
3-(2-Aminoethyl)-1H-indole-5,7-diol, 5,7-DHT hydrobromide, 5,7-DiHT hydrobromide, 7-Hydroxyserotonin hydrobromide
About This Item
Recommended Products
Assay
≥94% (HPLC)
form
powder
storage condition
desiccated
color
white to dark brown
solubility
H2O: 2 mg/mL, clear
storage temp.
2-8°C
SMILES string
NCCC1=CNC2=C(O)C=C(O)C=C21.[H]Br
InChI
1S/C10H12N2O2/c11-2-1-6-5-12-10-8(6)3-7(13)4-9(10)14/h3-5,12-14H,1-2,11H2
InChI key
LXWHQTNFZDTKBH-UHFFFAOYSA-N
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service