Skip to Content
Merck
All Photos(1)

Documents

84713

Sigma-Aldrich

Carbon disulfide

puriss., low in benzene, ≥99.5% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CS2
CAS Number:
Molecular Weight:
76.14
Beilstein:
1098293
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:

vapor density

2.67 (vs air)

Quality Level

vapor pressure

5.83 psi

grade

puriss.

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

212 °F

quality

low in benzene

expl. lim.

50 %

impurities

≤1 ppm benzene
≤1 ppm toluene

refractive index

n20/D 1.627 (lit.)
n20/D 1.627

bp

46 °C (lit.)

mp

−112-−111 °C (lit.)

solubility

water: soluble 2.9 g/L at 20 °C

density

1.263 g/mL at 20 °C
1.266 g/mL at 25 °C (lit.)

SMILES string

S=C=S

InChI

1S/CS2/c2-1-3

InChI key

QGJOPFRUJISHPQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1

Target Organs

Peripheral nervous system,Central nervous system,Cardio-vascular system,Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-22.0 °F - closed cup

Flash Point(C)

-30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global sources, lifetimes and mass balances of carbonyl sulfide (OCS) and carbon disulfide (CS2) in the earth's atmosphere.
Khalil MAK and Rasmussen RA.
Atmospheric Environment, 18(9), 1805-1813 (1984)
The mass budgets of carbonyl sulfide, dimethyl sulfide, carbon disulfide and hydrogen sulfide.
Watts SF.
Atmospheric Environment, 34(5), 761-779 (2000)
C60: a new form of carbon.
Kratschmer W, et al.
Nature, 347(6291), 354-358 (1990)
Charles-André Fustin et al.
Langmuir : the ACS journal of surfaces and colloids, 22(15), 6690-6695 (2006-07-13)
We report on a simple procedure to tune the hydrophilicity of hybrid gold nanoparticles. The nanoparticles have been prepared in the core of a poly(ethylene glycol)-block-poly(epsilon-caprolactone) (PEG-b-PCL) five-arm star block copolymer. A hydrophilic corona was then added to these hybrid
Strongly electron deficient sulfonyldithioformate based RAFT agents for hetero Diels-Alder conjugation: Computational design and experimental evaluation.
Nebhani L, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 47(22), 6053-6053 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service