Merck
  • Tuning the hydrophilicity of gold nanoparticles templated in star block copolymers.
Tuning the hydrophilicity of gold nanoparticles templated in star block copolymers.
Langmuir : the ACS journal of surfaces and colloids (2006-07-13)
Charles-André Fustin, Catheline Colard, Mariam Filali, Pierre Guillet, Anne-Sophie Duwez, Michael A R Meier, Ulrich S Schubert, Jean-François Gohy
ABSTRACT
We report on a simple procedure to tune the hydrophilicity of hybrid gold nanoparticles. The nanoparticles have been prepared in the core of a poly(ethylene glycol)-block-poly(epsilon-caprolactone) (PEG-b-PCL) five-arm star block copolymer. A hydrophilic corona was then added to these hybrid gold nanoparticles by direct chemisorption of trithiocarbonate-containing poly(acrylic acid) chains. These polymers were synthesized by RAFT polymerization with a trithiocarbonate as the chain-transfer agent. The efficiency of the grafting was evidenced by TEM, AFM, and DLS and by the successful transfer of these nanoparticles from organic solvent to water.
Sigma-Aldrich
Carbon disulfide, ACS reagent, ≥99.9%
Pricing and availability is not currently available.
Sigma-Aldrich
Carbon disulfide, suitable for IR spectroscopy, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Carbon disulfide, suitable for HPLC, ≥99.9%
Pricing and availability is not currently available.