Skip to Content
Merck
All Photos(1)

Key Documents

73879

Supelco

Piperonylbutoxide-(butyl-d9)

PESTANAL®, analytical standard

Synonym(s):

2-[2-(Butoxy-d9)ethoxy]ethyl (6-propylpiperonyl) ether, 4,5-Methylenedioxy-2-propylbenzyldiethyleneglycol butyl-d9 ether, Piperonylbutoxide-d9

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
R$2,822.00

R$2,822.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
R$2,822.00

About This Item

Empirical Formula (Hill Notation):
C19D9H21O5
CAS Number:
Molecular Weight:
347.49
MDL number:
UNSPSC Code:
77101502
NACRES:
NA.24

R$2,822.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

Related Categories

General description

Piperonylbutoxide-(butyl-d9) is an isotope labeled analog of the insecticide synergist piperonyl butoxide, wherein the butoxy protons are replaced by deuterium.[1]

Application

Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis, potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical behavior and determination of the insecticide synergist piperonyl butoxide
Coomber DC, et al.
Analytical Chemistry, 69(5), 898-903 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service