Skip to Content
Merck
All Photos(1)

Key Documents

90047

Supelco

Chlorpyrifos-(diethyl-d10)

PESTANAL®, analytical standard

Synonym(s):

O,O-Di(ethyl-d5) O-(3,5,6-trichloropyridin-2-yl) thiophosphate, Chlorpyriphos-d10

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$2,014.00

R$2,014.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$2,014.00

About This Item

Empirical Formula (Hill Notation):
C9D10HCl3NO3PS
CAS Number:
Molecular Weight:
360.65
MDL number:
UNSPSC Code:
77101502
NACRES:
NA.24

R$2,014.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

isotopic purity

≥95 atom % D

product line

PESTANAL®

Assay

≥97.5% (GC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

[P](=S)(OC(C([2H])([2H])[2H])([2H])[2H])(OC(C([2H])([2H])[2H])([2H])[2H])Oc1nc(c(cc1Cl)Cl)Cl

InChI

1S/C9H11Cl3NO3PS/c1-3-14-17(18,15-4-2)16-9-7(11)5-6(10)8(12)13-9/h5H,3-4H2,1-2H3/i1D3,2D3,3D2,4D2

InChI key

SBPBAQFWLVIOKP-MWUKXHIBSA-N

Related Categories

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service