Skip to Content
Merck
All Photos(1)

Key Documents

72669

Supelco

Fructose standard for IC

TraceCERT®, 1000 mg/L fructose in water (nominal concentration)

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
R$1,210.00

R$1,210.00


Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
50 ML
R$1,210.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

R$1,210.00


Estimated to ship onJune 02, 2025


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

concentration

1000 mg/L fructose in water (nominal concentration)

technique(s)

ion chromatography: suitable

application(s)

environmental
food and beverages

format

single component solution

storage temp.

2-8°C

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Linkage

Visit the IC Portal to learn more

Preparation Note

For the preparation of this CRM only materials of highest purity were used.

Analysis Note

1000 mg/L fructose in water, prepared with fructose and water; filtered through a 0.2 μm membrane filter

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

In this article we demonstrate how well ion chromatography is suited as an analytical technique for sugar analysis e.g. in apple juice, milk, and dust samples.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service