Skip to Content
Merck
All Photos(1)

Key Documents

62850

Sigma-Aldrich

L-Lysine solution

purum, 50% in H2O

Synonym(s):

(S)-2,6-Diaminocaproic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14N2O2
CAS Number:
Molecular Weight:
146.19
Beilstein:
1722531
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

grade

purum

optical activity

[α]20/D +13.0±1°, c = 2% in 6 M HCl

concentration

50% in H2O

application(s)

peptide synthesis

SMILES string

NCCCC[C@H](N)C(O)=O

InChI

1S/C6H14N2O2/c7-4-2-1-3-5(8)6(9)10/h5H,1-4,7-8H2,(H,9,10)/t5-/m0/s1

InChI key

KDXKERNSBIXSRK-YFKPBYRVSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sofia Faraasen et al.
Pharmaceutical research, 20(2), 237-246 (2003-03-15)
The purpose of this study was to demonstrate specific receptor-mediated targeting of phagocytes by functional surface coatings of microparticles, shielding from nonspecific phagocytosis and allowing ligand-specific interactions via molecular recognition. Coatings of the comb polymer poly(L-lysine)-g-poly(ethylene glycol) (PLL-g-PEG) were investigated
Lubin Jiang et al.
Nature, 499(7457), 223-227 (2013-07-05)
The variant antigen Plasmodium falciparum erythrocyte membrane protein 1 (PfEMP1), which is expressed on the surface of P. falciparum-infected red blood cells, is a critical virulence factor for malaria. Each parasite has 60 antigenically distinct var genes that each code
Sebastian Schröder et al.
Molecular cell, 52(3), 314-324 (2013-11-12)
Lysine acetylation regulates transcription by targeting histones and nonhistone proteins. Here we report that the central regulator of transcription, RNA polymerase II, is subject to acetylation in mammalian cells. Acetylation occurs at eight lysines within the C-terminal domain (CTD) of
Haig A Eskandarian et al.
Science (New York, N.Y.), 341(6145), 1238858-1238858 (2013-08-03)
Pathogens dramatically affect host cell transcription programs for their own profit during infection, but in most cases, the underlying mechanisms remain elusive. We found that during infection with the bacterium Listeria monocytogenes, the host deacetylase sirtuin 2 (SIRT2) translocates to
Nicholas I Proellocks et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 28(7), 3103-3113 (2014-04-08)
The genomes of malaria parasites (Plasmodium spp.) contain a family of genes encoding proteins with a Plasmodium helical interspersed subtelomeric (PHIST) domain, most of which are predicted to be exported into the parasite-infected human red blood cell (iRBC). Here, using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service