Merck
  • Acetylation of RNA polymerase II regulates growth-factor-induced gene transcription in mammalian cells.
Acetylation of RNA polymerase II regulates growth-factor-induced gene transcription in mammalian cells.
Molecular cell (2013-11-12)
Sebastian Schröder, Eva Herker, Friederike Itzen, Daniel He, Sean Thomas, Daniel A Gilchrist, Katrin Kaehlcke, Sungyoo Cho, Katherine S Pollard, John A Capra, Martina Schnölzer, Philip A Cole, Matthias Geyer, Benoit G Bruneau, Karen Adelman, Melanie Ott
ABSTRACT
Lysine acetylation regulates transcription by targeting histones and nonhistone proteins. Here we report that the central regulator of transcription, RNA polymerase II, is subject to acetylation in mammalian cells. Acetylation occurs at eight lysines within the C-terminal domain (CTD) of the largest polymerase subunit and is mediated by p300/KAT3B. CTD acetylation is specifically enriched downstream of the transcription start sites of polymerase-occupied genes genome-wide, indicating a role in early stages of transcription initiation or elongation. Mutation of lysines or p300 inhibitor treatment causes the loss of epidermal growth-factor-induced expression of c-Fos and Egr2, immediate-early genes with promoter-proximally paused polymerases, but does not affect expression or polymerase occupancy at housekeeping genes. Our studies identify acetylation as a new modification of the mammalian RNA polymerase II required for the induction of growth factor response genes.
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Pricing and availability is not currently available.
Supelco
L-Lysine Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Pricing and availability is not currently available.
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
ESGRO® Recombinant Mouse LIF Protein, ESGRO Leukemia Inhibitory Factor (LIF) supplement for mouse ES cell culture. Each vial contains 10^7 units/ml.
Pricing and availability is not currently available.
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Pricing and availability is not currently available.
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Lysine acetate salt, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Pricing and availability is not currently available.