Skip to Content
Merck
All Photos(2)

Documents

55443

Supelco

Fenofibrate

analytical standard

Synonym(s):

2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoic acid isopropyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21ClO4
CAS Number:
Molecular Weight:
360.83
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.3% water

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CC(C)OC(=O)C(C)(C)Oc1ccc(cc1)C(=O)c2ccc(Cl)cc2

InChI

1S/C20H21ClO4/c1-13(2)24-19(23)20(3,4)25-17-11-7-15(8-12-17)18(22)14-5-9-16(21)10-6-14/h5-13H,1-4H3

InChI key

YMTINGFKWWXKFG-UHFFFAOYSA-N

Gene Information

human ... PPARA(5465)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2 Oral

Target Organs

Liver

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samuel Rommelaere et al.
PloS one, 9(8), e104925-e104925 (2014-08-21)
Liver is a major regulator of lipid metabolism and adaptation to fasting, a process involving PPARalpha activation. We recently showed that the Vnn1 gene is a PPARalpha target gene in liver and that release of the Vanin-1 pantetheinase in serum
Pascale Massin et al.
Ophthalmic epidemiology, 21(5), 307-317 (2014-08-19)
Fenofibrate reduced progression of diabetic retinopathy in two large randomized studies. The effect of 135 mg fenofibric acid on diabetic macular edema (DME) was evaluated in subjects with existing DME. In this double-blind, randomized, placebo-controlled study, 110 subjects with DME not
Michel P Hermans
Diabetes & vascular disease research, 8(3), 180-189 (2011-05-18)
Microvascular complications are common in type 2 diabetes in primary care. Intensified management of glycaemia or blood pressure had little effect on microvascular complication rates in recent large trials (ADVANCE, VADT, ACCORD). In 2005, the FIELD study demonstrated a significant
Kate McKeage et al.
Drugs, 71(14), 1917-1946 (2011-09-29)
Fenofibrate is a fibric acid derivative indicated for the treatment of severe hypertriglyceridaemia and mixed dyslipidaemia in patients who have not responded to nonpharmacological therapies. The lipid-modifying effects of fenofibrate are mediated by the activation of peroxisome proliferator-activated receptor-α. Fenofibrate
Wen Huang et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 34(4), 646-653 (2014-01-16)
The strategies to protect against the disrupted blood-brain barrier (BBB) in HIV-1 infection are not well developed. Therefore, we investigated the potential of peroxisome proliferator-activated receptor (PPAR) agonists to prevent enhanced BBB permeability induced by HIV-1-specific protein Tat. Exposure to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service