52083
Thayer-Martin Supplement II
suitable for microbiology
Synonym(s):
Vitamino Growth Supplement
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
sterility
sterile
Quality Level
form
solid
shelf life
limited shelf life, expiry date on the label
application(s)
clinical testing
environmental
food and beverages
microbiology
storage temp.
2-8°C
suitability
Neisseria spp.
Related Categories
Application
A chemically defined growth supplement recommended for cultivation of Neisseria species.
Components
Composition (per vial; sufficient for 500 mL medium)
Part I (Cat. No.90541):
Vitamin B12 0.1mg; L-Glutamine 100mg; Adenine sulphate 10mg; Guanine hydrochloride 0.3mg; p-Amino benzoic acid (PABP) 0.13mg; L-Cystein 11mg; NAD (Coenzyme I) 2.5mg; Cocarboxylase 1mg; Ferric nitrate 0.2mg; Thiamine hydrochloride 0.03mg; Cysteine hydrochloride 259mg; Dextrose 1g
Part II (Rehydrating fluid; Cat. No. 18661):
Dextrose 1g; Distilled water 10ml
Part I (Cat. No.90541):
Vitamin B12 0.1mg; L-Glutamine 100mg; Adenine sulphate 10mg; Guanine hydrochloride 0.3mg; p-Amino benzoic acid (PABP) 0.13mg; L-Cystein 11mg; NAD (Coenzyme I) 2.5mg; Cocarboxylase 1mg; Ferric nitrate 0.2mg; Thiamine hydrochloride 0.03mg; Cysteine hydrochloride 259mg; Dextrose 1g
Part II (Rehydrating fluid; Cat. No. 18661):
Dextrose 1g; Distilled water 10ml
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service