Skip to Content
Merck
All Photos(1)

Documents

44051

Millipore

Thayer-Martin Supplement I

suitable for microbiology

Synonym(s):

V.C.N.T. Supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171614
NACRES:
NA.74

sterility

sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing
environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Neisseria spp.

Related Categories

Application

An antibiotic supplement recommended for the selective isolation of Neisseria gonorrhoeae and Neisseria meningitidis.

Components

composition (per vial, sufficient for 500 mL medium):
vancomycin 1.5 mg
colistin methane sulphonate 3.75 mg
trimethoprim 2.5 mg
nyastin 6′250 units

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service