Skip to Content
Merck
All Photos(1)

Key Documents

46573

Supelco

Estrone

VETRANAL®, analytical standard

Synonym(s):

1,3,5(10)-Estratrien-3-ol-17-one, 3-Hydroxy-1,3,5(10)-estratrien-17-one, Folliculin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22O2
CAS Number:
Molecular Weight:
270.37
Beilstein:
1915077
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

258-260 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1CCC2=O

InChI

1S/C18H22O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-16,19H,2,4,6-9H2,1H3/t14-,15-,16+,18+/m1/s1

InChI key

DNXHEGUUPJUMQT-CBZIJGRNSA-N

Gene Information

human ... ESR1(2099)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annamaria Lilienkampf et al.
Journal of medicinal chemistry, 52(21), 6660-6671 (2009-10-15)
Many breast tumors are hormone-dependent, and estrogens, especially estradiol (E2), have a pivotal role in their growth and development. 17beta-Hydroxysteroid dehydrogenase type 1 (17beta-HSD1) is a key enzyme in the biosynthesis of female sex steroids, catalyzing the NADPH-dependent reduction of
Emma M Ferguson et al.
Water research, 47(4), 1604-1615 (2013-01-12)
Estuaries are often the final repositories for aquatic pollutants but how estuarine hydrology influences the availability of marine- and freshwater-derived pollutants is not well understood, particularly for micro-pollutants such as endocrine disrupting chemicals. To address this knowledge gap, this study
Shanshan Zhao et al.
Breast cancer research : BCR, 16(2), R30-R30 (2014-03-29)
Paradoxically, a breast cancer risk reduction with conjugated equine estrogens (CEE) and a risk elevation with CEE plus medroxyprogesterone acetate (CEE + MPA) were observed in the Women's Health Initiative (WHI) randomized controlled trials. The effects of hormone therapy on serum sex
Yoko Urata et al.
The Journal of clinical endocrinology and metabolism, 98(4), 1583-1590 (2013-03-02)
Endometriosis is a chronic inflammatory disease in which immune response and production of estrogen in endometriotic tissues are involved in the development of the disease. Prostaglandin E2 (PGE2) stimulates aromatase (P450arom) expression in endometrioma stromal cells (ESCs) and increases the
Juilee Rege et al.
The Journal of clinical endocrinology and metabolism, 98(3), 1182-1188 (2013-02-07)
A broad analysis of adrenal gland-derived 19-carbon (C19) steroids has not been reported. This is the first study that uses liquid chromatography-tandem mass spectrometry to quantify 9 C19 steroids (androgens and their precursors), estrone, and estradiol in the adrenal vein

Articles

Milli-Q® ultrapure water systems deliver water free of organic contaminants , including hormones present in a lab's tap water, making it suitable for the most sensitive LC-MS/MS analyses.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service