Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Sterility
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing