Skip to Content
Merck
All Photos(1)

Key Documents

435589

Sigma-Aldrich

Sulfuric acid

reagent grade, 95-98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

reagent grade

vapor density

<0.3 (25 °C, vs air)

vapor pressure

1 mmHg ( 146 °C)

description

Nominally 95-98% H2SO4

Assay

95-98%

bp

~290 °C (lit.)

density

1.840 g/mL at 25 °C (lit.)

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amy C Keller et al.
Journal of ethnopharmacology, 121(2), 248-254 (2008-11-26)
Costus spicatus Sw. (Costaceae) is a prominent medicinal herb used by Dominicans in the Dominican Republic and the United States for the treatment of diabetes, a growing epidemic in the Hispanic community. An ethnobotanical survey of the Dominican community in
Chengpao Cao et al.
Molecules (Basel, Switzerland), 18(2), 1613-1625 (2013-01-30)
An improved synthesis of multifunctionalized pyrrolo[2,3,4-kl]acridine derivatives with different substituted patterns using silica sulfuric acid (SSA) as a heterogeneous catalyst under microwave irradiation conditions was developed. The reaction could be conducted by using readily available and inexpensive substrates within short
X H Zhao et al.
Environmental technology, 34(1-4), 263-268 (2013-03-28)
The purpose of this study was to develop an efficient and, possibly, a practically operated methodology to recover phosphorus (P) from P-saturated dewatered aluminium sludge cakes (DASC) after the DASC have been beneficially reused as constructed wetlands substrate for P-rich
Masoud Nasr-Esfahani et al.
Journal of nanoscience and nanotechnology, 13(7), 5004-5011 (2013-08-02)
Vanadatesulfuric acid (VSA), as a novel and heterogeneous nanorod catalyst, was used for an efficient synthesis of tetrahydrobenzo[b]pyrans using an aldehydes, 1,3-cyclohexanediones or beta-ketoester and malononitrile in C2H5OH/H2O mixture under reflux conditions. VSA is prepared via the reaction of sodium
Young Hoon Jung et al.
Bioresource technology, 132, 109-114 (2013-02-12)
Dilute sulfuric acid pretreatment of oil palm empty fruit bunches (EFB) followed by the whole slurry fermentation of the pretreated EFB slurry was investigated. The optimized pretreatment conditions were at 1% (w/v) sulfuric acid with 3 min ramping to 190

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service