Merck
  • Phosphorus recovery as AlPO4 from beneficially reused aluminium sludge arising from water treatment.
Phosphorus recovery as AlPO4 from beneficially reused aluminium sludge arising from water treatment.
Environmental technology (2013-03-28)
X H Zhao, Y Q Zhao, P Kearney
ABSTRACT
The purpose of this study was to develop an efficient and, possibly, a practically operated methodology to recover phosphorus (P) from P-saturated dewatered aluminium sludge cakes (DASC) after the DASC have been beneficially reused as constructed wetlands substrate for P-rich wastewater treatment. A three-step procedure of 1) P extraction by H2SO4, 2) decolorization of extraction leachate via H2O2 oxidation, and 3) AlPO4 precipitation by pH adjustment, has been explored. The optimal conditions to form the precipitates of AlPO4 were determined, with 97% of P and 99% of Al being recovered. The obtained compounds were identified by XRD, FTIR and SEM analyses. Although the purity, structure, characteristics and production control of the compounds are worthy of further investigation, this study provides a showcase of a 'closed loop' regarding the beneficial reuse of a 'waste' and the recovery of useful elements after the reuse.
Product Description
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Aluminum phosphate monobasic, purum, ≥95.0% (gravimetric)
Sigma-Aldrich
Aluminum phosphate, 99.99% trace metals basis
Sigma-Aldrich
Aluminum phosphate, reagent grade
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Aluminum phosphate monobasic solution, ~50% in H2O