Skip to Content
Merck
All Photos(1)

Documents

40163

Supelco

Dimethyldioctadecylammonium bromide

Selectophore, ≥99.0%

Synonym(s):

Distearyldimethylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)17]2N(Br)(CH3)2
CAS Number:
Molecular Weight:
630.95
Beilstein:
3580323
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

description

cationic

product line

Selectophore

Assay

≥99.0% (T)
≥99.0%

mol wt

630.95 g/mol

mp

~160 °C

SMILES string

CCCCCCCCCCCCCCCCCC[N+](CCCCCCCCCCCCCCCCCC)(C)C.[Br-]

InChI

1S/C38H80N.BrH/c1-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-39(3,4)38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-2;/h5-38H2,1-4H3;1H/q+1;/p-1

InChI key

PSLWZOIUBRXAQW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Dimethyldioctadecylammonium bromide acts as a cationic surfactant.

Application

It was used in treating microporous polypropylene membrane filters, in order to modify and create charged surface which was used to filter nanoparticles.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Filtration of nanoparticles with dimethyldioctadecylammonium bromide treated microporous polypropylene filters.
Kang, Peter K., and Dinesh O. Shah
Langmuir, 13, 1820-1826 (1997)
Randip Kaur et al.
Journal of controlled release : official journal of the Controlled Release Society, 164(3), 331-337 (2012-07-18)
The mechanism behind the immunostimulatory effect of the cationic liposomal vaccine adjuvant dimethyldioctadecylammonium and trehalose 6,6'-dibehenate (DDA:TDB) has been linked to the ability of these cationic vesicles to promote a depot after administration, with the liposomal adjuvant and the antigen
Zhi Rao et al.
Colloids and surfaces. B, Biointerfaces, 97, 248-253 (2012-05-23)
In order to confirm that the cholesterol end groups of cholesterol-end capped polyethylene glycol really insert into the liposome bilayer and investigate how the incorporation affects the microenvironment of liposome bilayer, two kinds of molecular probes, namely Nile Red and
Paolo Sabatino et al.
The journal of physical chemistry. B, 116(30), 9269-9276 (2012-06-27)
The interaction between phenol molecules, both in their undissociated and dissociated states, and cationic dioctadecyl dimethylammonium chloride (DODAC) vesicles were thoroughly investigated using NMR techniques. In particular, diffusion and relaxation measurements, combined with the two sites Kärger model, were used
Yu-Qian Wang et al.
International immunopharmacology, 17(1), 9-17 (2013-04-30)
As an ideal tumor antigen, survivin has been widely used for tumor immunotherapy. Nevertheless, no effective protein vaccine targeting survivin has been reported, which may be due to its poor ability to induce cellular immunity. Thus, a suitable immunoadjuvant and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service