Skip to Content
Merck
All Photos(1)

Key Documents

34466

Supelco

Didecyldimethylammonium chloride

analytical standard

Synonym(s):

N-Decyl-N,N-dimethyl-1-decanaminium chloride, DDAC

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
R$521.00

R$521.00


Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
250 MG
R$521.00

About This Item

Empirical Formula (Hill Notation):
C22H48ClN
CAS Number:
Molecular Weight:
362.08
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

R$521.00


Estimated to ship onJune 01, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

SMILES string

C[N+](C)(CCCCCCCCCC)CCCCCCCCCC.[Cl-]

InChI

1S/C22H48N.ClH/c1-5-7-9-11-13-15-17-19-21-23(3,4)22-20-18-16-14-12-10-8-6-2;/h5-22H2,1-4H3;1H/q+1;/p-1

InChI key

RUPBZQFQVRMKDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alice Roedel et al.
Microorganisms, 8(4) (2020-04-16)
We investigated the suitability of a newly developed biocide susceptibility test system based on microtiter plates containing vacuum dried biocides as a fast and reliable screening method. The evaluated substances included the cationic biocides benzalkonium chloride (BAC), chlorhexidine dihydrochloride (CHX)
Hana Malenovská
Food and environmental virology, 12(4), 361-366 (2020-10-16)
The global SARS-CoV-2 pandemic dictates that anti-contagion strategies should become matters of essential routine in everyday life. Fomite transference is one of the routes of transmission that has been considered for this virus. However, the risks associated with contaminated surfaces
Çiğdem Arpa et al.
Journal of analytical methods in chemistry, 2018, 8951028-8951028 (2018-08-30)
Ultrasound-assisted dispersive liquid-liquid microextraction method based on solidification of floating organic drop and back-extraction (UA-DLLME-SFO-BE) technique was proposed for preconcentration of lead ions. In this technique, two SFODME steps are applied in sequence. The classical SFODME was applied as the
H Uzuner et al.
The Journal of hospital infection, 100(3), e30-e36 (2018-05-25)
Hand antiseptics are of great importance in the prevention of nosocomial infections. Due to the frequent occurrence of adenoviral epidemic keratoconjunctivitis outbreaks in the study hospital, it was necessary to investigate the efficiency of alcohol-based solutions on adenoviruses. To investigate
Robin S Wegh et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 34(11), 1935-1947 (2017-08-08)
A non-targeted workflow is reported for the isolation and identification of antimicrobial active compounds using bioassay-directed screening and LC coupled to high-resolution MS. Suspect samples are extracted using a generic protocol and fractionated using two different LC conditions (A and

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service