Skip to Content
Merck
All Photos(1)

Key Documents

34800

Sigma-Aldrich

2,6-Di-tert-butylphenol

purum, ≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]2C6H3OH
CAS Number:
Molecular Weight:
206.32
Beilstein:
1841887
EC Number:
MDL number:
UNSPSC Code:
12352100

vapor pressure

<0.01 mmHg ( 20 °C)

grade

purum

Assay

≥98.0% (GC)

bp

253 °C (lit.)

mp

34-37 °C (lit.)
34-37 °C

SMILES string

CC(C)(C)c1cccc(c1O)C(C)(C)C

InChI

1S/C14H22O/c1-13(2,3)10-8-7-9-11(12(10)15)14(4,5)6/h7-9,15H,1-6H3

InChI key

DKCPKDPYUFEZCP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

227.3 °F - closed cup

Flash Point(C)

108.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jörg Ahrens et al.
Anesthesia and analgesia, 99(1), 91-96 (2004-07-30)
The anesthetic propofol (2,6 diisopropylphenol) mediates some of its effects by activating inhibitory chloride currents in the lower brainstem and spinal cord. The effects comprise direct activation of gamma-aminobutyric acid-A and glycine receptors in the absence of the natural agonist
Synthesis and biological evaluation of 2,6-di-tert-butylphenol hydrazones as 5-lipoxygenase inhibitors.
A M Cuadro et al.
Bioorganic & medicinal chemistry, 6(2), 173-180 (1998-04-21)
Juan Ruiz et al.
Bioorganic & medicinal chemistry, 11(19), 4207-4216 (2003-09-03)
The dual or selective ability of 24 derived mono- and 2,6-di-tert-butylphenols (DTBP) to act as inhibitors of cyclooxygenase (COX) and/or 5-lipoxygenase (LOX) enzymes is investigated. Firstly, we explored the conformational variability of the compounds. It is found that dual inhibitors
Zhen-wei Fang et al.
Huan jing ke xue= Huanjing kexue, 25(3), 98-101 (2004-08-26)
A degrading bacterial strain F-3-4 for 2,6-Di-tert-butylphenol (2,6-DTBP) was isolated from biofilm in acrylic fiber wastewater treatment structures. By acclimation, its capacity for degradation of 2,6-DTBP was enhanced by 26%. It was identified as Alcaligenes sp. according to morphological, physiological
Jörg Ahrens et al.
Pharmacology, 83(2), 95-98 (2008-12-10)
Modulation of inhibitory synaptic transmission within the central nervous system contributes considerably to the anaesthetic effects of propofol and its analogues in vivo. We have studied the effects of the non-anaesthetic propofol analogue 2,6-di-tert-butylphenol on rat alpha(1)beta(2)gamma(2) GABA(A) receptors expressed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service