Skip to Content
Merck
All Photos(3)

Documents

D48400

Sigma-Aldrich

2,6-Di-tert-butylphenol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]2C6H3OH
CAS Number:
Molecular Weight:
206.32
Beilstein:
1841887
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

99%

form

crystals

bp

253 °C (lit.)

mp

34-37 °C (lit.)

SMILES string

CC(C)(C)c1cccc(c1O)C(C)(C)C

InChI

1S/C14H22O/c1-13(2,3)10-8-7-9-11(12(10)15)14(4,5)6/h7-9,15H,1-6H3

InChI key

DKCPKDPYUFEZCP-UHFFFAOYSA-N

Gene Information

human ... GABRA1(2554)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

227.3 °F - closed cup

Flash Point(C)

108.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anouar Hafiane et al.
The Canadian journal of cardiology, 35(6), 770-781 (2019-06-04)
Small peptides based on the C-terminal domain of apo E have recently been proposed as ATP-binding cassette transporter A1 (ABCA1) agonist with therapeutic potential. Previous work has shown that a novel synthetic peptide, CS-6253, acts synergistically with apolipoprotein A-I or
Caitlin E Cornell et al.
Biophysical journal, 113(6), 1200-1211 (2017-08-13)
A persistent challenge in membrane biophysics has been to quantitatively predict how membrane physical properties change upon addition of new amphiphiles (e.g., lipids, alcohols, peptides, or proteins) in order to assess whether the changes are large enough to plausibly result
G Haeseler et al.
European journal of anaesthesiology, 20(3), 220-224 (2003-03-26)
Propofol is a phenol derivative (2,6 di-isopropylphenol) with a unique effect profile including activating effects on GABA(A) and blocking effects on voltage-operated sodium channels. If the substituents in the 2- and the 6-positions are replaced by tert-butyl groups, the resulting
Stanley J Stachelek et al.
Journal of biomedical materials research. Part A, 82(4), 1004-1011 (2007-03-21)
Polyurethane cardiovascular implants are subject to oxidation initiated surface degradation, which is mediated by monocyte-derived macrophages (MDM); this often leads to surface cracking and device failure. The present studies examined the hypothesis that covalently attaching antioxidant, di-tert-butylphenol (DBP), to the
A W Girotti et al.
Lipids, 22(6), 401-408 (1987-06-01)
The effects of singlet oxygen- and oxygen radical-induced lipid peroxidation on cell membrane integrity were compared, using the human erythrocyte ghost as a model system. Resealed ghosts underwent lipid peroxidation and lysis (release of trapped glucose-6-P) when irradiated in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service