Skip to Content
Merck
All Photos(2)

Key Documents

32965

Supelco

Bentazon-d7

PESTANAL®, analytical standard

Synonym(s):

3-Isopropyl-d7-1H-2,1,3-benzothiadiazin-4(3H)-one 2,2-dioxide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
R$4,743.00

R$4,743.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
R$4,743.00

About This Item

Empirical Formula (Hill Notation):
C10D7H5N2O3S
CAS Number:
Molecular Weight:
247.32
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

R$4,743.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

mass shift

M+7

SMILES string

[2H]C([2H])([2H])C([2H])(N1C(=O)c2ccccc2NS1(=O)=O)C([2H])([2H])[2H]

InChI

1S/C10H12N2O3S/c1-7(2)12-10(13)8-5-3-4-6-9(8)11-16(12,14)15/h3-7,11H,1-2H3/i1D3,2D3,7D

InChI key

ZOMSMJKLGFBRBS-QXMYYZBZSA-N

General description

Bentazon-d7 is a deuterated form of bentazon, typically used for environmental and food residue analysis.[1]

Application

Bentazon-d7 may be used as an internal standard for the determination of bentazon in shellfish by ultra-high performance liquid chromatography-high resolution mass spectrometry (UHPLC-HRMS).[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fast analysis of relevant contaminants mixture in commercial shellfish
Alvarez-Mu?oz D, et al.
Talanta (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service