Skip to Content
Merck
All Photos(2)

Key Documents

32186

Supelco

Oxolinic acid-(ethyl-d5)

VETRANAL®, analytical standard

Synonym(s):

5-(Ethyl-d5)-8-oxo-5,8-dihydro-1,3-dioxolo[4,5-g]quinoline-7-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
R$5,131.00

R$5,131.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
R$5,131.00

About This Item

Empirical Formula (Hill Notation):
C13D5H6NO5
CAS Number:
Molecular Weight:
266.26
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

R$5,131.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

storage temp.

2-8°C

SMILES string

[n]1(c2c([c](c(c1)C(=O)O)=O)cc3c(c2)OCO3)C(C([2H])([2H])[2H])([2H])[2H]

InChI

1S/C13H11NO5/c1-2-14-5-8(13(16)17)12(15)7-3-10-11(4-9(7)14)19-6-18-10/h3-5H,2,6H2,1H3,(H,16,17)/i1D3,2D2

InChI key

KYGZCKSPAKDVKC-ZBJDZAJPSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service