Skip to Content
Merck
All Photos(1)

Key Documents

31678

Supelco

Cloquintocet-mexyl

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
R$716.00

R$716.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
R$716.00

About This Item

Empirical Formula (Hill Notation):
C18H22ClNO3
CAS Number:
Molecular Weight:
335.83
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

R$716.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCCCCC(C)OC(=O)COc1ccc(Cl)c2cccnc12

InChI

1S/C18H22ClNO3/c1-3-4-5-7-13(2)23-17(21)12-22-16-10-9-15(19)14-8-6-11-20-18(14)16/h6,8-11,13H,3-5,7,12H2,1-2H3

InChI key

COYBRKAVBMYYSF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cloquintocet-mexyl is a colourless crystalline herbicide safener. It is categorized as Class III toxin.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Urinary bladder

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

384.8 °F - closed cup

Flash Point(C)

196 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kalyani Paranjape
The Pesticide Encyclopedia, 110-110 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service