![Cloquintocet-mexyl PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/774/342/af4307b3-107b-42be-a3f8-1b2b95676c4c/640/af4307b3-107b-42be-a3f8-1b2b95676c4c.png)
Cloquintocet-mexyl
![Cloquintocet-mexyl PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/774/342/af4307b3-107b-42be-a3f8-1b2b95676c4c/640/af4307b3-107b-42be-a3f8-1b2b95676c4c.png)
Empirical Formula (Hill Notation):
C18H22ClNO3
CAS Number:
Molecular Weight:
335.83
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing